Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Whatman™ Polyester Drain Discs

Polyester drain disc, PE, membrane filter accessories, 47 mm, 100 µm
  CA11013-696
 :   Cytiva
 :  231100
Whatman™ Polyester Drain Discs
  CA11013-696
 :  Cytiva
Whatman™ Polyester Drain Discs
Whatman™ Polyester Drain Discs

Polyester drain disc, PE, membrane filter accessories, 47 mm, 100 µm

 
$258.81
   
Pack of 100
 

The polyester drain disc is binder free and has a thickness of 100 µm.

  • Flat surface to eliminate filter tearing or rupturing
  • Used as a separator between membrane layers in serial stack filtration applications

This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.

 
  • Ø:
    47 mm
  • Désignation:
    Polyester drain disc, PE, membrane filter accessories
  • Épaisseur:
    100 µm