Accessories for General Purpose Heating Circulators, Grant Instruments
Replacement bridge-plate, fits T optima on ST12, ST18, ST26 and ST38 tanks
89260-880
Supplier #: T-BTL
Accessories for General Purpose Heating Circulators, Grant Instruments
89260-880
- Désignation:Replacement bridge-plate, fits T optima on ST12, ST18, ST26 and ST38 tanks
Accessories
- Description
- 89260-854Clamp to attach Thermostat to Independent Tank; allows independent use of Optima™ Immersion Thermostat, models T100, TC120, TX150, and TXF200Each - $268.61
- Description
- Description