Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Electrophoresis Running Buffers, G-Biosciences

MOPS/ SDS Running Buffer (20X), 500 mL
Catalog # CA95043-526
Supplier:   G-Biosciences
 :  786-532
Electrophoresis Running Buffers, G-Biosciences
Catalog # CA95043-526
Supplier:  G-Biosciences
CAS Number:  
Electrophoresis Running Buffers, G-Biosciences

MOPS/ SDS Running Buffer (20X), 500 mL

CAS Number:  
Price
$361.18
Log in pour la disponibilité
Each (1l)
 
Product Details & Documents

G-Biosciences offers many electrophoresis running buffers including: TAE Running Buffer [50X], TBE Running Buffer [10X], TBE-Urea Sample Buffer, MOPS Running Buffer [10X], and MOPS/ SDS Running Buffer [20X]

  • The TAE Running Buffer [50X] contains 2M Tris-Acetate and 0.05M EDTA (at a pH of 8.3)
  • The TBE Running Buffer [10X] contains 0.9M Tris-borate and 0.01M EDTA (at a pH of 8.3)
  • The MOPS Running Buffer [20X] contains 200 mM MOPS (at a pH of 7.0), 80 mM Sodium acetate and 10 mM EDTA

G-Biosciences' TBE-Urea Sample Buffer is a loading buffer used to resolve single-stranded DNA oligos or RNA into sharp, distinct bands. The TBE-Urea Sample Buffer is for the analysis and purification of products ranging from 20-800 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and Northern blot analysis.

These include the SDS-PAGE Sample Loading Buffer, SDS-PAGE Running Buffer, Native Sample Loading Buffer, Tris Glycine Native Gel Running Buffer, Tricine Sample Buffer, Tris-Tricine, Tris-Tricine-SDS, MES SDS Running Buffer, MOPS/ SDS Running Buffer, IEF Anode Buffer, IEF Cathode Buffer, Zymogram Sample Buffer, Zymogram Renature Buffer, Zymogram Development Buffer and the Discontinuous Buffer System.

Spécifications

Spécifications

  • Désignation:
    MOPS/ SDS Running Buffer (20X)
  • Taille:
    500 mL

Spécifications