FirstChoice™ Blocking Agents, G-Biosciences
FirstChoice™ Blocking Agents, G-Biosciences
FirstChoice™ Blocking Buffer-TBS
Catalog # CA89167-866
Supplier:   G-Biosciences
Supplier #:  786-666
FirstChoice™ Blocking Agents, G-Biosciences
Catalog # CA89167-866
Supplier:  G-Biosciences
CAS Number:  
FirstChoice™ Blocking Agents, G-Biosciences
FirstChoice™ Blocking Buffer-TBS
CAS Number:  
Price
$448.58
Log in pour la disponibilité
Each (500ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
G-Biosciences' FirstChoice™ Blocking Buffer is ideal as a first choice for optimization of new assays and systems, or when determining the optimal blocking buffer for elimination of non-specific binding sites in ELISA, blotting, immunohistochemistry and other applications.
G-Biosciences' FirstChoice™ Blocking Buffer is a proprietary protein formulation that offers greater versatility and lack of cross-reactivity.
G-Biosciences' FirstChoice™ Blocking Buffers are compatible with antibodies and avidin/biotin based systems. The results show high signal to background ratios. This ready-to-use blocking agent is animal serum free and biotin free.
G-Biosciences' FirstChoice™ Blocking Buffers are conveniently supplied in widely used TBS (Tris-buffered saline at pH 7.5) and PBS (phosphate-buffered saline at pH 7.5) buffers as well as in separate formulations containing Tween® 20 for improving blocking efficiencies.
Ordering information: Accessories available.
Spécifications
Spécifications
  • Désignation:
    FirstChoice™ Blocking Buffer-TBS