Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Superior™ Blocking Buffer, Protein Blocking Agent in Multiple Formats, G-Biosciences | Avantor

Superior™ Blocking Buffer, Protein Blocking Agent in Multiple Formats, G-Biosciences

Superior™ Blocking Buffer in PBST
Catalog # CA89167-856
Supplier:   G-Biosciences
Fournisseur #:  786-661
Superior™ Blocking Buffer, Protein Blocking Agent in Multiple Formats, G-Biosciences
Catalog # CA89167-856
Supplier:  G-Biosciences
CAS Number:  
Superior™ Blocking Buffer, Protein Blocking Agent in Multiple Formats, G-Biosciences

Superior™ Blocking Buffer in PBST

CAS Number:  
Price
$473.39
Log in pour la disponibilité
Each (500ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

G-Biosciences' Superior™ Blocking Buffer is an enhanced blocking agent available in multiple formats

G-Biosciences' Superior™ Blocking Buffer is ideal for a high signal to background ratio in most systems and has a rapid blocking time of about two minutes for ELISA. Superior™ Blocking Buffer uses a non-serum protein and does not contain biotin or other animal source proteins that interfere with immuno-complexes. Superior™ Blocking Buffer is suitable for assays that use avidin/streptavidin systems.
G-Biosciences' Superior™ Blocking Buffer for Precipitating Substrate is a modification of the original product that has been optimized for use in blotting protocols that use precipitating substrates such as G-Biosciences' femtoCHROMO™ chromogenic detection systems, TMB (3, 3’, 5, 5’-Tetramethylbenzidine), BCIP (5-Bromo-4-Chloro-3’-Indolyphosphate p-Toluidine Salt) and NBT (Nitro-Blue Tetrazolium Chloride) substrates. The Superior™ Blocking Buffer for Precipitating Substrate is not suitable for ELISA or immunohistochemistry staining.
G-Biosciences' Superior™ Blocking Buffer is available in multiple ready-to-use liquid formats: in TBS, PBS, TBS with 0.05% Tween® 20 detergent, or PBS with 0.05% Tween® 20. Also available is a convenient dry form that is stable at room temperature. Each dry buffer pack makes 200ml Superior™ Blocking Buffer (user is supplied with 5 packs).

Superior™ Blocking Buffer contains a proprietary antigenically non-determinant protein for blocking Western blot membranes, blocking and storage of ELISA plates, and blocking prior to immunohistochemistry staining.

Ordering information: Accessories available.

Spécifications

Spécifications

  • Désignation:
    Superior™ Blocking Buffer in PBST

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.