Swiss Tweezer; Precision, Electron Microscopy Sciences | Avantor
Swiss Tweezer; Precision, Electron Microscopy Sciences
Style 5AG
Catalog # 102098-952
Fournisseur #:  78170-5AG
Swiss Tweezer; Precision, Electron Microscopy Sciences
Catalog # 102098-952
Supplier:  
Swiss Tweezer; Precision, Electron Microscopy Sciences
Swiss Tweezer; Precision, Electron Microscopy Sciences
Style 5AG
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
  • Tweezers engineered for a NON-SLIP GRIP
  • Light weight –prevents hand fatigue
  • Sensuous touch and functional design
  • Ultra-fine precision tips
  • Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.
Spécifications
  • Désignation:
    Style 5AG