Swiss Tweezer; Precision, Electron Microscopy Sciences | Avantor
Swiss Tweezer; Precision, Electron Microscopy Sciences
Style Tweeze Model 2G
Catalog # 102098-944
Fournisseur #:  78170-2G
Swiss Tweezer; Precision, Electron Microscopy Sciences
Catalog # 102098-944
Swiss Tweezer; Precision, Electron Microscopy Sciences
Swiss Tweezer; Precision, Electron Microscopy Sciences
Style Tweeze Model 2G
Price
$70.99
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
  • Tweezers engineered for a NON-SLIP GRIP
  • Light weight –prevents hand fatigue
  • Sensuous touch and functional design
  • Ultra-fine precision tips
  • Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.
Spécifications
  • Désignation:
    Style Tweeze Model 2G