Labeling Tape, Electron Microscopy Sciences
Green, 0.75" x 2160"
  102098-768
 :  77628-G
Labeling Tape, Electron Microscopy Sciences
  102098-768
Labeling Tape, Electron Microscopy Sciences
Labeling Tape, Electron Microscopy Sciences
Green, 0.75" x 2160"
$109.43
Each Roll of 1
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
  • Color:
    Green
  • Taille:
    0.75" x 2160"