Labeling Tape, Electron Microscopy Sciences
Yellow, 0.75" x 500"
Catalog # 102098-764
 :  77627-Y
Labeling Tape, Electron Microscopy Sciences
Catalog # 102098-764
Labeling Tape, Electron Microscopy Sciences
Labeling Tape, Electron Microscopy Sciences
Yellow, 0.75" x 500"
Price
37,41 $
Log in pour la disponibilité
Each Roll of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
Spécifications
  • Color:
    Yellow
  • Taille:
    0.75" x 500"