Labeling Tape, Electron Microscopy Sciences
White, 0.75" x 500"
  102098-762
 :  77627-W
Labeling Tape, Electron Microscopy Sciences
  102098-762
Labeling Tape, Electron Microscopy Sciences
Labeling Tape, Electron Microscopy Sciences
White, 0.75" x 500"
$32.49
Each Roll of 2
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
  • Color:
    White
  • Taille:
    0.75" x 500"