Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, 32 x 13 mm
  102098-752
 :  77626-10
Write Protect Tape, Electron Microscopy Sciences
  102098-752
Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, 32 x 13 mm
87,34 $
Each Roll of 1,000
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers.
  • Will not dry out, peel or fall off
  • Withstand temperatures ranging from –196 to 150 °C
The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 and 1.5 mL tubes.
  • Désignation:
    Write Protect Tape
  • Dimensions:
    32 x 13 mm