Specialty Labels, Electron Microscopy Sciences
Product Details & Documents
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Spécifications
- Color:Dark green
- Désignation:Tough-Spots
- Legend:Bullseye
- Diameter:7/16"