Thermal Transfer Labels, Electron Microscopy Sciences | Avantor
Thermal Transfer Labels, Electron Microscopy Sciences
Thermal transfer labels, General purpose, 38 x 19 mm
Catalog # 102098-680
Fournisseur #:  77592-87
Thermal Transfer Labels, Electron Microscopy Sciences
Catalog # 102098-680
Thermal Transfer Labels, Electron Microscopy Sciences
Thermal Transfer Labels, Electron Microscopy Sciences
Thermal transfer labels, General purpose, 38 x 19 mm
Price
$871.17
Log in pour la disponibilité
Each Roll of 4,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Thermal Transfer Cryo-Tags are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers. Pre-cut, peel-off labels designed for use in thermal transfer printers. Print popular barcode formats for fast and easy sample identification. Works with all thermal transfer printers.
  • Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Specially coated for use with a resin ribbon that provides the best performance under extreme conditions
  • Resists Degradation from scuffing, chemicals, moisture and wide temperature fluctuations
  • Thermal Transfer Labels for Storage in Temperatures From −196°C to 80°C
  • Print as little as one label at a time, eliminating label waste and saving time and money
  • Print crisp, clear, professional quality labels
Spécifications
  • Pour l'utilisation avec:
    General purpose
  • Désignation:
    Thermal transfer labels
  • Dimensions:
    38 x 19 mm