Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
White, 54.8 m (60 yds.), 12.7 mm (½")
  102098-444
 :  77011-W1
Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
  102098-444
Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
White, 54.8 m (60 yds.), 12.7 mm (½")
$55.48
Each
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250°F). It may be used as well down to -73°C.
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core. Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
  • Largeur:
    12.7 mm (½")
  • Color:
    White
  • Longueur:
    54.8 m (60 yds.)