Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Product Details & Documents
Spécifications
- Désignation:Hitachi M4 12 divisions
- Hauteur:6.0 mm (¼")
- Matière:Aluminum
- Head Diameter:32 mm (1¼")