Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 32 mm (1¼"), 6.0 mm (¼")
  102098-190
 :  75635-70
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
  102098-190
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 32 mm (1¼"), 6.0 mm (¼")
$119.56
Each Pack of 10
  • Désignation:
    Hitachi M4
  • Hauteur:
    6.0 mm (¼")
  • Matière:
    Aluminum
  • Head Diameter:
    32 mm (1¼")