Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 25 mm (1"), 10 mm (⅜")
  102098-186
 :  75635-50
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
  102098-186
 :  
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 25 mm (1"), 10 mm (⅜")
  • Désignation:
    Hitachi M4
  • Hauteur:
    10 mm (⅜")
  • Matière:
    Aluminum
  • Head Diameter:
    25 mm (1")