Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 15 mm (⅝"), 10 mm (⅜")
  102098-182
 :  75635-30
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
  102098-182
 :  
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
Hitachi M4, Aluminum, 15 mm (⅝"), 10 mm (⅜")
  • Désignation:
    Hitachi M4
  • Hauteur:
    10 mm (⅜")
  • Matière:
    Aluminum
  • Head Diameter:
    15 mm (⅝")