Histo-Mark Pen, Electron Microscopy Sciences
Product Details & Documents
The fine-line Histo-Mark pen is designed to work like a pen.
- Can be used on any surface
- Marking quality remains consistent throughout the life of the ink barrel
The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene.
Spécifications
- Point Type:Fine
- Color:Black
- Désignation:Histo-Mark pen