Disposable Scalpel Sets, Electron Microscopy Sciences
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
: Packed 10 each /box.
- Désignation:Scalpel Handle Set #15