Disposable Scalpel Sets, Electron Microscopy Sciences
Disposable Scalpel Sets, Electron Microscopy Sciences
Scalpel Handle Set #11
Catalog # 102097-854
Supplier #:  72052-11
Disposable Scalpel Sets, Electron Microscopy Sciences
Catalog # 102097-854
Disposable Scalpel Sets, Electron Microscopy Sciences
Scalpel Handle Set #11
Price
$29.85
Log in for availability
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Packaging: Packed 10 each /box.
Specifications
  • Désignation:
    Scalpel Handle Set #11