Feather® Safeshield™ Scalpels, Electron Microscopy Sciences | Avantor
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Blade #22
  102097-812
Fournisseur #:  72043-22
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
  102097-812
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Blade #22
$113.14
Each Pack of 10
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures
  • Heat-treated
  • Comfortable hand fit
  • Retractable shield
  • Color-coded blade size identification
They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Packaging: Individually packaged, 10 per box.
  • Désignation:
    Feather® Safeshield™ Blade #22