Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Product Details & Documents
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures
- Heat-treated
- Comfortable hand fit
- Retractable shield
- Color-coded blade size identification
They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
: Individually packaged, 10 per box.
Spécifications
- Désignation:Feather® Safesheild™ Blade #15