Feather® Safeshield™ Scalpels, Electron Microscopy Sciences | Avantor
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Blade #11
Catalog # 102097-800
Fournisseur #:  72043-11
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Catalog # 102097-800
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Feather® Safeshield™ Blade #11
Price
$100.51
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures
  • Heat-treated
  • Comfortable hand fit
  • Retractable shield
  • Color-coded blade size identification
They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Packaging: Individually packaged, 10 per box.
Spécifications
  • Désignation:
    Feather® Safeshield™ Blade #11