Wash-Off Labels, Electron Microscopy Sciences | Avantor
Wash-Off Labels, Electron Microscopy Sciences
Plain, 4×1"
Catalog # 102097-662
Fournisseur #:  71826-20
Wash-Off Labels, Electron Microscopy Sciences
Catalog # 102097-662
Supplier:  
Wash-Off Labels, Electron Microscopy Sciences
Wash-Off Labels, Electron Microscopy Sciences
Plain, 4×1"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rinses away leaving no adhesive residue. Labels dissolve under running water in less than 30 seconds. Labels Wash-Off in 1 rinse cycle at any water temperature.
  • Autoclavable labels resist exposure to steam at 121 °C, a build-up of liquid water on the label surface will dissolve the material
  • Temperature-resistant
  • Accepts laser printing and other markings containers
Environmentally Safe, 100% biodegradable. Long term storage best between 32 to 70 °F (0 to 21 °C).
Certifications: FDA Compliant.
Spécifications
  • Désignation:
    Plain
  • Dimensions:
    4×1"