Pop-Up Slide Holder, Electron Microscopy Sciences
Product Details & Documents
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".
Spécifications
- Désignation:30 Slide Pop-Up Slide Folder