Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences | Avantor
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray
Catalog # 102097-586
Fournisseur #:  71503-10
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Catalog # 102097-586
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray
Price
$179.70
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This durable ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1”).
Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable. Measures 11 7/8” L x 7 ”W x ½”H (300 x 195 x 12 mm).
Spécifications
  • Désignation:
    Hard Plastic Microscope Slide Tray