Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray
  102097-584
 :  71503-01
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
  102097-584
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Hard Plastic Microscope Slide Tray
$20.51
Each
This durable ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1”).
Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable. Measures 11 7/8” L x 7 ”W x ½”H (300 x 195 x 12 mm).
  • Désignation:
    Hard Plastic Microscope Slide Tray