Slide Box-Foam, Electron Microscopy Sciences | Avantor
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 100, Black
Catalog # 102097-576
Fournisseur #:  71478-18
Slide Box-Foam, Electron Microscopy Sciences
Catalog # 102097-576
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 100, Black
Price
$24.94
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
  • Holds 25, 50, or 100 slides
  • Foam backing
  • Secure lock
  • Blue, green, red, yellow, white, black, and pink
Spécifications
  • Capacité:
    100
  • Color:
    Black
  • Désignation:
    Slide Box