Slide Box-Foam, Electron Microscopy Sciences | Avantor
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 25, White
Catalog # 102097-550
Fournisseur #:  71478-05
Slide Box-Foam, Electron Microscopy Sciences
Catalog # 102097-550
Supplier:  
Slide Box-Foam, Electron Microscopy Sciences
Slide Box-Foam, Electron Microscopy Sciences
Slide Box, 25, White
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR SLIDEBOX25PLACE CORK WHITEE Catalog #82003-426
Le produit 82003-426 est similaire à 102097-550 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
  • Holds 25, 50, or 100 slides
  • Foam backing
  • Secure lock
  • Blue, green, red, yellow, white, black, and pink
Spécifications
  • Capacité:
    25
  • Color:
    White
  • Désignation:
    Slide Box