Staining Dishes, Electron Microscopy Sciences
Staining dish only, 120×122×110 mm
  102097-518
 :  71424-D
Staining Dishes, Electron Microscopy Sciences
  102097-518
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Staining dish only, 120×122×110 mm
$218.90
Each
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and stainless steel lids.
  • A variety of sizes are available
    Lids and dishes are available separately
Ideal for immersing slides in several solutions.
  • Désignation:
    Staining dish only
  • Dimensions:
    120×122×110 mm