EasyDip™ Slide Staining System, Electron Microscopy Sciences
The EasyDip™ Slide Staining System has two components: a square staining jar and a 12-position vertical slide staining rack. Jars can be loosely joined to each other laterally using the stainless steel holder making sure they are kept in the same order when moved around on the lab counter. The jars are available in five different colors to help better identity contents or applications.
The Staining Jar being made from acetal plastic will not break like most glass jars do. It will resist attacks from most staining reagents including alcohol and xylenes but not phenols. The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80 mL. Ideal for frozen sections, small jobs and special processes. Dimensions: 64×76×92 mm (21/2×3×35/8").
The EasyDip™ Slide Staining Rack will hold up to 12 slides of 25×75 mm or 26×76 mm (3×1") with a thickness of 1.0 to 1.2 mm. These racks are safe at temperatures up to 120 °C. The lid completely covers the EasyDip™ Slide Staining Jar to minimize spill and evaporation. The handle is permanently attached to the rack for easy insertion or removal of the slide. The base of the rack is placed in the vertical position to ensure the slides are in place, and are rotated sideways for easy slide removal. Dimensions: 57 x 64 x 95 mm (21/4×21/2×33/4").
EasyDip™ Stainless Steel Holder is designed to hold six EasyDip™ staining jars together. Measures: 425 x 102 x 38 mm (163/4L×4W×11/2"H).
: For your convenience, we offer EasyDip™ Slide Staining Kit. Kit consists of one Staining Jar and one Staining Rack, color white.
- Color:Pink
- Désignation:Slide Staining Jar