Cryogenic Grid Box, Electron Microscopy Sciences
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
- Four diamond shaped slots
- Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
- Box only fits 71166-20 (Grid box square with lid)
- Available with lid, pin-type lid, or without lid
- Color:Blue
- Désignation:Grid Box Round without Lid