Cryogenic Grid Box, Electron Microscopy Sciences | Avantor
Cryogenic Grid Box, Electron Microscopy Sciences
Grid Box Square with Lid, Blue
Catalog # 102097-444
Fournisseur #:  71166-20
Cryogenic Grid Box, Electron Microscopy Sciences
Catalog # 102097-444
Cryogenic Grid Box, Electron Microscopy Sciences
Cryogenic Grid Box, Electron Microscopy Sciences
Grid Box Square with Lid, Blue
Price
$35.84
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
  • Four diamond shaped slots
  • Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
  • Box only fits 71166-20 (Grid box square with lid)
  • Available with lid, pin-type lid, or without lid
Spécifications
  • Color:
    Blue
  • Désignation:
    Grid Box Square with Lid