EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Product Details & Documents
Modules have 24 slots labeled with letters from A-X where the grids can be stored and rotated for easy access as well as 3 additional cavities for block storage .
Available with and without a unique identification number.
Spécifications
- Désignation:Dial A Grid-N-Block storage box with unique number