EMS Grid Box, Electron Microscopy Sciences
EMS Grid Box, Electron Microscopy Sciences
50 Grid Storage Box
Catalog # 102097-418
Supplier #:  71146-01
EMS Grid Box, Electron Microscopy Sciences
Catalog # 102097-418
EMS Grid Box, Electron Microscopy Sciences
EMS Grid Box, Electron Microscopy Sciences
50 Grid Storage Box
Price
$29.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The EMS50 and EMS100 TEM grid storage boxes are used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage for standard grids that are 3.05 mm in diameter.
  • Dimensions for both boxes are: 75×55×6.5 mm
  • Anti static treated
The EMS 50 and EMS100 capacity inexpensive grid storage boxes. The box has a simple number/letter combination printed on the side of the body.
Specifications
  • Désignation:
    50 Grid Storage Box