Polyethylene Disposable Gloves, Electron Microscopy Sciences
Product Details & Documents
Low cost, offers good protection to hands from chemical contamination.
- Excellent Sensitivity
- Powderless/sulfur-free
- Ambidextrous
- 1¾ mil thickness
Packaging: 100 gloves per roll. Comes in a Dispenser Box.
Documents
Spécifications
- Taille du gant:M
- Cat. No.:102097-324
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous