Parafilm Dispenser, Electron Microscopy Sciences | Avantor
Parafilm Dispenser, Electron Microscopy Sciences
Replacement Blades for Dispenser
Catalog # 102097-224
Fournisseur #:  70993-04
Parafilm Dispenser, Electron Microscopy Sciences
Catalog # 102097-224
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Replacement Blades for Dispenser
Price
$27.14
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company
Spécifications
  • Désignation:
    Replacement Blades for Dispenser