Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser Red
Catalog # 102097-222
Supplier #:  70993-03
Parafilm Dispenser, Electron Microscopy Sciences
Catalog # 102097-222
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser Red
Price
$183.91
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company
Spécifications
  • Désignation:
    Parafilm Dispenser Red