Parafilm Dispenser, Electron Microscopy Sciences
Product Details & Documents
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
- Holds one or two 2" rolls, or one 4" roll
- Size (W x H x D): 120 x 156 x 171 mm
- Parafilm is a registered trademark of American can company
Spécifications
- Désignation:Parafilm Dispenser Blue