Checker Plus® pH Tester, Electron Microscopy Sciences
  102097-184
Checker Plus® pH Tester, Electron Microscopy Sciences
  102097-184
 :  
Checker Plus® pH Tester, Electron Microscopy Sciences
Checker Plus® pH Tester, Electron Microscopy Sciences
The Checker Plus electronic pH tester supplies you with fast and accurate readings on a large LCD. The electrode can be replaced, allowing the tester to last for years.
  • Large and easy-to-read LCD
  • High accuracy with 0.01 pH resolution
  • Automatic, one or two points calibration
  • Battery life: 1,000 hours of continuous use
Modern Substitute for Litmus Paper with Replaceable Electrode. The extended family of Checker® pH testers now includes standard gel-filled electrodes with built-in BNC or screw type connectors.
Checker Plus is great for environmental, field and laboratory testing. Also suitable for checking electrodes in pipes and tanks and various laboratory applications.
Accessories
  •  
    Description
    For Use With
    Connector
  • 100493-012
    HI 1271 replacement electrode
    Model 70699-02
    Screw type
    Each - $57.34