Fluor Adhesive Chamber, Electron Microscopy Sciences
22 mm x 60 mm, 1, 140 - 165 µl, 0.12 mm
  102096-640
 :  70328-55
Fluor Adhesive Chamber, Electron Microscopy Sciences
  102096-640
Fluor Adhesive Chamber, Electron Microscopy Sciences
Fluor Adhesive Chamber, Electron Microscopy Sciences
22 mm x 60 mm, 1, 140 - 165 µl, 0.12 mm
$1,007.99
Each Pack of 100
Each package includes illustrated instructions for use, applicator, and 200 adhesive port seals
These new microarray chambers are designed specially for compatibility with Cy5® or Alexa Fluor 647® direct labeled DNA probes for hybridization assays. These chambers as well as the original HybriWells may also be used with all other fluorochrome labeled probes.
 : Additional seal tabs are sold separately: Adesive Port Seal Tabs.
  • Profondeur:
    0.12 mm
  • Nb de puits:
    1
  • Volume:
    140 - 165 µl
  • Taille:
    22 mm x 60 mm