BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
  102096-556
 :  70010-B
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
  102096-556
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
$99.28
Each Pack of 500
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.
  • Ø:
    8 mm
  • Désignation:
    Embedding Capsules Size 00
Accessories
  •  
    Description
  • 100490-718
    Capsule Holder for Size 00
    Each - $232.67
  •  
    Description
  • 100492-862
    Capsule Holder, Clear
    Each - $40.76
  •  
    Description
  • 100492-864
    Capsule Holder for Size 00
    Each - $49.04
  •  
    Description
  • 100488-950
    Embedding Capsule Holder
    Each - $123.24
  •  
    Description
  • 100488-954
    Embedding Capsule Holder
    Each - $283.55
  •  
    Description
  • 100492-884
    Embedding Capsule Holder
    Each - $35.65