BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences | Avantor
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
Catalog # 102096-550
Fournisseur #:  70000-B
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Catalog # 102096-550
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Capsules Size 00, 8 mm
Price
$21.59
Log in pour la disponibilité
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.
Spécifications
  • Ø:
    8 mm
  • Désignation:
    Embedding Capsules Size 00
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Capsule Holder for Size 00
    Capsule Holder for Size 00
    100490-718
    Log in pour la disponibilité
    Each - $232.67
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Capsule Holder, Clear
    Capsule Holder, Clear
    100492-862
    Log in pour la disponibilité
    Each - $40.76
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Capsule Holder for Size 00
    Capsule Holder for Size 00
    100492-864
    Log in pour la disponibilité
    Each - $49.04
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100488-950
    Log in pour la disponibilité
    Each - $123.24
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100488-954
    Log in pour la disponibilité
    Each - $283.55
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Embedding Capsule Holder
    Embedding Capsule Holder
    100492-884
    Log in pour la disponibilité
    Each - $35.65