BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
  102096-536
Supplier #:  69910-10
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
  102096-536
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
$192.44
Each Pack of 1,000
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
  • Désignation:
    Embedding Capsules, Size 3
Accessories
  •  
    Description
  • 103301-920
    BEEM Capsule Holder White for Size 3
    Each - $31.32
  •  
    Description
  • 103302-262
    BEEM Capsule Holder White for Size 3
    Each - $180.15
  •  
    Description
  • 103305-110
    BEEM Capsule Holder White for Size 3
    Each - $337.62