BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
  102096-534
 :  69910-05
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
  102096-534
 :  
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Capsules, Size 3
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
  • Désignation:
    Embedding Capsules, Size 3
Accessories
  •  
    Description
  • 103301-920
    BEEM Capsule Holder White for Size 3
    Each - $31.32
  •  
    Description
  • 103302-262
    BEEM Capsule Holder White for Size 3
    Each - $180.15
  •  
    Description
  • 103305-110
    BEEM Capsule Holder White for Size 3
    Each - $337.62