BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Product Details & Documents
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Spécifications
- Désignation:Embedding Capsules, Size 3
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- BEEM Capsule Holder White for Size 3
Informations sur le produit
- Catalog #AvailabilityPrice
- BEEM Capsule Holder White for Size 3
Informations sur le produit
- Catalog #AvailabilityPrice
- BEEM Capsule Holder White for Size 3