Accu-Punch, Electron Microscopy Sciences
Accu-Punch, Electron Microscopy Sciences
Assorted One of Each Size
Catalog # 102096-488
Supplier #:  69038-AS
Accu-Punch, Electron Microscopy Sciences
Catalog # 102096-488
Accu-Punch, Electron Microscopy Sciences
Accu-Punch, Electron Microscopy Sciences
Assorted One of Each Size
Price
$332.62
Log in for availability
Each Pack of 14
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Accu-Punch is a sterile, disposable skin biopsy punch.
  • Always sharp and sterile
  • Ergonomic design
  • Ribbed for maximum control
  • Available in 14 different sizes in a dispenser box
It is the only disposable punch made in the USA.
Specifications
  • Désignation:
    Assorted One of Each Size