Accu-Punch, Electron Microscopy Sciences
Punch, 3.5 mm
  102096-480
 :  69038-35
Accu-Punch, Electron Microscopy Sciences
  102096-480
Accu-Punch, Electron Microscopy Sciences
Accu-Punch, Electron Microscopy Sciences
Punch, 3.5 mm
$322.39
Each Pack of 25
The Accu-Punch is a sterile, disposable skin biopsy punch.
  • Always sharp and sterile
  • Ergonomic design
  • Ribbed for maximum control
  • Available in 14 different sizes in a dispenser box
It is the only disposable punch made in the USA.
  • Désignation:
    Punch, 3.5 mm