Accu-Punch, Electron Microscopy Sciences
Punch, 6.0 mm
Catalog # 102096-454
 :  69038-06-50
Accu-Punch, Electron Microscopy Sciences
Catalog # 102096-454
Accu-Punch, Electron Microscopy Sciences
Accu-Punch, Electron Microscopy Sciences
Punch, 6.0 mm
Price
$655.63
Log in pour la disponibilité
Each Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Accu-Punch is a sterile, disposable skin biopsy punch.
  • Always sharp and sterile
  • Ergonomic design
  • Ribbed for maximum control
  • Available in 14 different sizes in a dispenser box
It is the only disposable punch made in the USA.
Spécifications
  • Désignation:
    Punch, 6.0 mm